The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021551.4 | NP_067526.2 | 1 - 2 | 271 | 102 |
GenBank mRNA | AK007059.1 | - | 1 - 2 | 216 | 102 |
BC005727.1 | - | 1 - 2 | 233 | 102 |