Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Olfm3
Entrez Gene ID
229759
Gene Name
olfactomedin 3
Gene Aliases
B230206G02Rik
Chromosome Location
Chr.3: 114904078 - 115125764 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286750.1 NP_001273679.1 1 - 2 73 71
GenBank mRNA BB628879.1 - 1 - 2 74 71

View Full Product Details