Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Synpr
Entrez Gene ID
72003
Gene Name
synaptoporin
Gene Aliases
1500003F20Rik, SPO
Chromosome Location
Chr.14: 13284780 - 13615469 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028052.4 NP_082328.3 1 - 2 225 70
GenBank mRNA AK005132.1 - 1 - 2 146 70
AK078223.1 - 1 - 2 226 70
BC026512.1 - 1 - 2 172 70

View Full Product Details