Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Tmem56
- Entrez Gene ID
-
99887
- Gene Name
- transmembrane protein 56
- Gene Aliases
-
4930577M16Rik, AU018104, AW109744, C730036B01Rik
- Chromosome Location
-
Chr.3: 121202008 - 121283108 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK016295.1 |
- |
1 - 2 |
202 |
65 |
View Full Product Details