The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026945.1 | NP_081221.1 | 4 - 5 | 407 | 61 |
GenBank mRNA | AK007397.1 | - | 4 - 5 | 403 | 61 |
AK028114.1 | - | 4 - 5 | 408 | 61 | |
BC130016.1 | - | 4 - 5 | 376 | 61 |