Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cma1
Entrez Gene ID
17228
Gene Name
chymase 1, mast cell
Gene Aliases
MMCP-5, Mcp-5, Mcp5, Mcpt5
Chromosome Location
Chr.14: 55941451 - 55944661 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010780.2 NP_034910.1 2 - 3 331 117
GenBank mRNA BC139454.1 - 2 - 3 320 117
BC139457.1 - 2 - 3 320 117
BC139759.1 - 2 - 3 275 117
M68898.1 - 2 - 3 207 117
M73759.1 - 2 - 3 230 117
X68805.1 - 2 - 3 332 117

View Full Product Details