Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Otud1
Entrez Gene ID
71198
Gene Name
OTU domain containing 1
Gene Aliases
4933428L19Rik
Chromosome Location
Chr.2: 19658062 - 19660590 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027715.1 NP_081991.1 1 - 1 1448 108
GenBank mRNA AK016972.1 - 1 - 1 755 108

View Full Product Details