Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Armc3
Entrez Gene ID
70882
Gene Name
armadillo repeat containing 3
Gene Aliases
4921513G22Rik
Chromosome Location
Chr.2: 19198865 - 19310243 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081083.2 NP_001074552.1 14 - 15 2073 92
NM_001271563.1 NP_001258492.1 14 - 15 2073 92
NM_001271564.1 NP_001258493.1 14 - 15 2073 92
NM_001271565.1 NP_001258494.1 14 - 15 2073 92
GenBank mRNA AK014880.1 - 14 - 15 1889 92
AK041744.1 - 14 - 15 1897 92
BC145004.1 - 14 - 15 1881 92
BC145005.1 - 14 - 15 1881 92
BC145013.1 - 14 - 15 1881 92
BC150688.1 - 14 - 15 1881 92

View Full Product Details