Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tbr1
Entrez Gene ID
21375
Gene Name
T-box brain gene 1
Gene Aliases
-
Chromosome Location
Chr.2: 61802734 - 61814114 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009322.3 NP_033348.2 1 - 2 946 91
GenBank mRNA BC052737.1 - 1 - 2 946 91
BC058399.1 - 1 - 2 850 91
U49251.1 - 1 - 2 786 91

View Full Product Details