Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Per1
Entrez Gene ID
18626
Gene Name
period circadian clock 1
Gene Aliases
Per, m-rigui, mPer1
Chromosome Location
Chr.11: 69095140 - 69109965 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159367.1 NP_001152839.1 16 - 17 2213 65
NM_011065.4 NP_035195.2 16 - 17 2228 65
GenBank mRNA AB002108.1 - 15 - 16 2042 65
AF022992.1 - 16 - 17 2267 65
AK081813.1 - 16 - 17 2226 65
AK148202.1 - 16 - 17 2220 65
AK154900.1 - 16 - 17 2209 65
AK172958.1 - 16 - 17 2213 65
BC039768.1 - 15 - 16 2113 65
BC091645.1 - 16 - 17 2215 65

View Full Product Details