The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033041.4 | NP_149030.2 | 2 - 3 | 160 | 94 |
GenBank mRNA | AB049065.1 | - | 2 - 3 | 139 | 94 |
AK050791.1 | - | 2 - 3 | 161 | 94 | |
BC134378.1 | - | 2 - 3 | 165 | 94 |