The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010452.3 | NP_034582.1 | 4 - 5 | 855 | 71 |
GenBank mRNA | BC096612.1 | - | 3 - 4 | 831 | 71 |
U56399.1 | - | 1 - 2 | 364 | 71 | |
Y11717.1 | - | 4 - 5 | 905 | 71 |