The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_180678.3 | NP_851009.2 | 15 - 16 | 1916 | 74 |
GenBank mRNA | AK012758.2 | - | 15 - 16 | 1911 | 74 |
AK146238.1 | - | 15 - 16 | 1893 | 74 | |
AK165857.1 | - | 15 - 16 | 1864 | 74 | |
AK182981.1 | - | 1 - 2 | 98 | 74 | |
AK198707.1 | - | 1 - 2 | 98 | 74 | |
AK198907.1 | - | 1 - 2 | 98 | 74 | |
BC021747.1 | - | 15 - 16 | 1897 | 74 |