The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001335.2 | NP_001001335.1 | 11 - 12 | 1460 | 98 |
NM_001164361.1 | NP_001157833.1 | 11 - 12 | 1555 | 98 | |
GenBank mRNA | AK078040.1 | - | 11 - 12 | 1556 | 98 |
AK170592.1 | - | 11 - 12 | 1320 | 98 | |
BC052360.1 | - | 11 - 12 | 1460 | 98 |