Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Camsap1
Entrez Gene ID
227634
Gene Name
calmodulin regulated spectrin-associated protein 1
Gene Aliases
9530003A05Rik, C77823, PRO2405
Chromosome Location
Chr.2: 25926838 - 25983452 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276359.1 NP_001263288.1 10 - 11 1740 71
NM_001276360.1 NP_001263289.1 11 - 12 1773 71
NM_001276361.1 NP_001263290.1 11 - 12 1800 71
GenBank mRNA AK035221.1 - 3 - 4 252 71
BC024715.2 - 10 - 11 1455 71
BC054553.1 - 11 - 12 1579 71

View Full Product Details