The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013460.4 | NP_038488.4 | 1 - 2 | 1206 | 66 |
GenBank mRNA | S80044.1 | - | 1 - 2 | 1206 | 66 |