The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001109761.2 | NP_001103231.1 | 19 - 20 | 2119 | 58 |
NM_001177799.1 | NP_001171270.1 | 19 - 20 | 2386 | 58 | |
NM_007601.3 | NP_031627.2 | 22 - 23 | 2662 | 58 | |
GenBank mRNA | AK049994.1 | - | 9 - 10 | 548 | 58 |
AK079673.1 | - | 4 - 5 | 338 | 58 | |
BC090661.1 | - | 19 - 20 | 2119 | 58 |