Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Map1a
Entrez Gene ID
17754
Gene Name
microtubule-associated protein 1 A
Gene Aliases
6330416M19Rik, AI853608, Mtap-1, Mtap1, Mtap1a, moth1
Chromosome Location
Chr.2: 121289600 - 121310832 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032393.2 NP_115769.1 2 - 3 373 75
GenBank mRNA AF182213.1 - 2 - 3 373 75

View Full Product Details