Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vmn1r55
Entrez Gene ID
384522
Gene Name
vomeronasal 1 receptor 55
Gene Aliases
Gm1422, Gm403, V1rd5
Chromosome Location
Chr.7: 5146490 - 5147422 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166706.1 NP_001160178.1 1 - 1 484 118

View Full Product Details