The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033369.3 | NP_001028541.1 | 6 - 7 | 938 | 71 |
NM_001111030.1 | NP_001104500.1 | 7 - 8 | 1305 | 71 | |
GenBank mRNA | AK049089.1 | - | 6 - 7 | 939 | 71 |
AK142396.1 | - | 7 - 8 | 1309 | 71 | |
BC028780.1 | - | 3 - 4 | 323 | 71 | |
BC145224.1 | - | 6 - 7 | 1101 | 71 |