Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ambra1
Entrez Gene ID
228361
Gene Name
autophagy/beclin 1 regulator 1
Gene Aliases
2310079H06Rik, A130023A14, AA474864, AV021921, D030051N19Rik, mKIAA1736
Chromosome Location
Chr.2: 91730111 - 91918849 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080754.1 NP_001074223.1 2 - 3 492 66
NM_172669.3 NP_766257.3 2 - 3 492 66
GenBank mRNA AK037516.1 - 2 - 3 449 66
AK083596.1 - 2 - 3 449 66
AK084557.1 - 2 - 3 473 66
AK148887.1 - 2 - 3 484 66
AK155513.1 - 2 - 3 455 66
AK169280.1 - 2 - 3 445 66
BC032293.1 - 2 - 3 415 66
BC054082.1 - 2 - 3 424 66

View Full Product Details