The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009105.4 | NP_033131.2 | 8 - 9 | 951 | 77 |
GenBank mRNA | AK009288.1 | - | 8 - 9 | 841 | 77 |
AK011857.1 | - | 8 - 9 | 844 | 77 | |
AK013076.1 | - | 4 - 5 | 422 | 77 | |
AK014749.1 | - | 3 - 4 | 294 | 77 | |
AK150011.1 | - | 8 - 9 | 819 | 77 | |
AK152655.1 | - | 8 - 9 | 831 | 77 | |
X63039.1 | - | 8 - 9 | 766 | 77 |