Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gpcpd1
Entrez Gene ID
74182
Gene Name
glycerophosphocholine phosphodiesterase GDE1 homolog (S. cerevisiae)
Gene Aliases
2310004G06Rik, 2310032D16Rik, AU015213, Gde5, Prei4, mKIAA1434
Chromosome Location
Chr.2: 132529082 - 132578282 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042672.1 NP_001036137.1 12 - 13 1362 127
NM_001291050.1 NP_001277979.1 11 - 12 1235 127
NM_001291051.1 NP_001277980.1 11 - 12 1265 127
NM_001291052.1 NP_001277981.1 11 - 12 1283 127
NM_027096.2 NP_081372.1 10 - 11 1159 127
NM_028802.3 NP_083078.3 12 - 13 1359 127
GenBank mRNA AK009563.1 - 5 - 6 578 127
AK030645.1 - 12 - 13 1313 127
AK031658.1 - 11 - 12 1238 127
AK049491.1 - 4 - 5 448 127
AK050318.1 - 12 - 13 1274 127
AK051728.1 - 4 - 5 422 127
AK141374.1 - 12 - 13 1249 127
AK163419.1 - 11 - 12 1206 127
AK165952.1 - 12 - 13 1324 127
AK166293.1 - 3 - 4 293 127
AK166454.1 - 11 - 12 1248 127
BC006887.1 - 11 - 12 1207 127
BC033408.1 - 10 - 11 1089 127

View Full Product Details