Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bdnf
Entrez Gene ID
12064
Gene Name
brain derived neurotrophic factor
Gene Aliases
-
Chromosome Location
Chr.2: 109674700 - 109727043 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001285416.1 NP_001272345.1 1 - 2 417 113
GenBank mRNA AY057910.1 - 1 - 2 417 113
EF125671.1 - 1 - 2 418 113
HM623886.1 - 1 - 2 157 113

View Full Product Details