Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lhfpl1
Entrez Gene ID
237091
Gene Name
lipoma HMGIC fusion partner-like 1
Gene Aliases
Lhfp
Chromosome Location
Chr.X: 145290359 - 145372304 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178358.3 NP_848135.1 3 - 4 803 68
GenBank mRNA AK142526.1 - 3 - 4 860 68
AY253666.1 - 3 - 4 859 68
BC051434.1 - 3 - 4 803 68

View Full Product Details