Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fbln7
Entrez Gene ID
70370
Gene Name
fibulin 7
Gene Aliases
1600015H20Rik, AI464334, TM14
Chromosome Location
Chr.2: 128863862 - 128897034 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024237.4 NP_077199.2 4 - 5 677 143
GenBank mRNA BC051550.1 - 2 - 3 233 143
BC095941.1 - 4 - 5 609 143

View Full Product Details