The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001110228.1 | NP_001103698.1 | 7 - 8 | 847 | 78 |
NM_001110229.1 | NP_001103699.1 | 8 - 9 | 934 | 78 | |
NM_001110230.1 | NP_001103700.1 | 8 - 9 | 934 | 78 | |
NM_001110231.1 | NP_001103701.1 | 10 - 11 | 909 | 78 | |
NM_001110232.1 | NP_001103702.1 | 9 - 10 | 920 | 78 | |
NM_001160292.1 | NP_001153764.1 | 8 - 9 | 934 | 78 | |
NM_001160293.1 | NP_001153765.1 | 7 - 8 | 847 | 78 | |
NM_010160.2 | NP_034290.2 | 7 - 8 | 847 | 78 | |
GenBank mRNA | AF090696.1 | - | 9 - 10 | 920 | 78 |
AF090697.1 | - | 7 - 8 | 820 | 78 | |
AK135166.1 | - | 7 - 8 | 799 | 78 | |
AK137292.1 | - | 8 - 9 | 875 | 78 | |
AK151818.1 | - | 6 - 7 | 705 | 78 | |
AK152628.1 | - | 6 - 7 | 705 | 78 | |
AK153267.1 | - | 6 - 7 | 705 | 78 | |
AK159996.1 | - | 8 - 9 | 939 | 78 | |
AK160861.1 | - | 10 - 11 | 910 | 78 | |
AK162651.1 | - | 5 - 6 | 4410 | 78 | |
AK164914.1 | - | 8 - 9 | 875 | 78 | |
BC026856.1 | - | 7 - 8 | 798 | 78 |