Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ankrd9
Entrez Gene ID
74251
Gene Name
ankyrin repeat domain 9
Gene Aliases
2500003O20Rik
Chromosome Location
Chr.12: 110975353 - 110979021 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175207.4 NP_780416.2 2 - 3 357 94
GenBank mRNA AK032641.1 - 2 - 3 358 94
BC023687.1 - 2 - 3 331 94

View Full Product Details