Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vmn1r65
Entrez Gene ID
81013
Gene Name
vomeronasal 1 receptor 65
Gene Aliases
V1rd6, V3R6
Chromosome Location
Chr.7: 6007750 - 6011010 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030738.2 NP_109663.1 1 - 2 158 60
GenBank mRNA AF324870.1 - 1 - 2 158 60

View Full Product Details