The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133204.2 | NP_573467.2 | 1 - 2 | 56 | 76 |
GenBank mRNA | AK139659.1 | - | 1 - 2 | 57 | 76 |