Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Amot
Entrez Gene ID
27494
Gene Name
angiomotin
Gene Aliases
CAG-2, D0Kist1, Sii6
Chromosome Location
Chr.X: 145446424 - 145505541 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290274.1 NP_001277203.1 7 - 8 2051 78
NM_153319.3 NP_695231.3 7 - 8 2051 78
GenBank mRNA AF461135.1 - 7 - 8 1455 78
AK129277.1 - 2 - 3 291 78
BC057638.1 - 7 - 8 1506 78
CK392840.1 - 4 - 5 497 78

View Full Product Details