The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029232.2 | NP_083508.1 | 1 - 2 | 122 | 116 |
GenBank mRNA | AK016214.1 | - | 1 - 2 | 113 | 116 |
BC050803.1 | - | 1 - 2 | 122 | 116 |