Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc38a11
Entrez Gene ID
320106
Gene Name
solute carrier family 38, member 11
Gene Aliases
9330158F14Rik
Chromosome Location
Chr.2: 65315799 - 65364394 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177074.2 NP_796048.2 5 - 6 545 138
GenBank mRNA AK043610.1 - 5 - 6 554 138
AK139966.1 - 5 - 6 495 138

View Full Product Details