The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013482.2 | NP_038510.2 | 18 - 19 | 2067 | 127 |
GenBank mRNA | BC053392.1 | - | 18 - 19 | 2003 | 127 |
L08967.1 | - | 18 - 19 | 2043 | 127 | |
L10627.1 | - | 18 - 19 | 2075 | 127 | |
L29788.1 | - | 18 - 19 | 2039 | 127 |