Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Armcx6
Entrez Gene ID
278097
Gene Name
armadillo repeat containing, X-linked 6
Gene Aliases
AW060994
Chromosome Location
Chr.X: 134748447 - 134756342 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007578.2 NP_001007579.1 3 - 4 255 77
GenBank mRNA AK148507.1 - 3 - 4 254 77
AK160766.1 - 3 - 4 256 77
BC027264.1 - 3 - 4 248 77
BY777182.1 - 3 - 4 235 77

View Full Product Details