Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cfp
Entrez Gene ID
18636
Gene Name
complement factor properdin
Gene Aliases
BCFG, Pfc
Chromosome Location
Chr.X: 20925535 - 20931525 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008823.3 NP_032849.2 6 - 7 996 82
GenBank mRNA AK150212.1 - 6 - 7 957 82
AK152782.1 - 6 - 7 947 82
AK154306.1 - 6 - 7 981 82
AK159291.1 - 6 - 7 997 82
AK159609.1 - 6 - 7 947 82
AK159853.1 - 6 - 7 947 82
AK171374.1 - 6 - 7 980 82
BC132369.1 - 6 - 7 980 82
BC138306.1 - 6 - 7 980 82
X12905.1 - 5 - 6 845 82

View Full Product Details