Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cfp
Entrez Gene ID
18636
Gene Name
complement factor properdin
Gene Aliases
BCFG, Pfc
Chromosome Location
Chr.X: 20925535 - 20931525 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008823.3 NP_032849.2 7 - 8 1185 79
GenBank mRNA AK150212.1 - 7 - 8 1146 79
AK152782.1 - 7 - 8 1136 79
AK154306.1 - 7 - 8 1170 79
AK159291.1 - 7 - 8 1186 79
AK171374.1 - 7 - 8 1169 79
BC132369.1 - 7 - 8 1169 79
BC138306.1 - 7 - 8 1169 79
X12905.1 - 6 - 7 1034 79

View Full Product Details