Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Clec16a
Entrez Gene ID
74374
Gene Name
C-type lectin domain family 16, member A
Gene Aliases
4932416N17Rik, curt, mKIAA0350
Chromosome Location
Chr.16: 10545339 - 10744878 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204229.1 NP_001191158.1 8 - 9 1178 85
NM_177562.5 NP_808230.2 9 - 10 1184 85
GenBank mRNA AK089327.1 - 8 - 9 1108 85
AK122255.1 - 9 - 10 1136 85
AK155314.1 - 8 - 9 1120 85
AK156927.1 - 8 - 9 1109 85
AK170754.1 - 8 - 9 1109 85
AK171134.1 - 8 - 9 1109 85
BC138907.1 - 9 - 10 1087 85
BC138909.1 - 9 - 10 1087 85

View Full Product Details