Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mzt2
Entrez Gene ID
72083
Gene Name
mitotic spindle organizing protein 2
Gene Aliases
2410018G20Rik, 2610001E06Rik, Fam128, Fam128b
Chromosome Location
Chr.16: 15848441 - 15863364 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029354.2 NP_083630.1 1 - 2 570 99
GenBank mRNA AK010544.1 - 1 - 2 571 99
AK011255.1 - 2 - 3 331 99
BC027550.1 - 1 - 2 495 99

View Full Product Details