Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Thap6
Entrez Gene ID
381650
Gene Name
THAP domain containing 6
Gene Aliases
4930534K21Rik, AI448650
Chromosome Location
Chr.5: 91962882 - 91972137 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_028429.1 1 - 2 638 101
GenBank mRNA AK015967.1 - 2 - 3 875 101
BC089377.1 - 1 - 2 187 101

View Full Product Details