The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008599.4 | NP_032625.2 | 3 - 4 | 330 | 135 |
GenBank mRNA | AK042017.1 | - | 3 - 4 | 332 | 135 |
AK042480.1 | - | 3 - 4 | 248 | 135 | |
AK156012.1 | - | 3 - 4 | 331 | 135 | |
AK156029.1 | - | 3 - 4 | 331 | 135 | |
AK156453.1 | - | 3 - 4 | 331 | 135 | |
AK157109.1 | - | 3 - 4 | 331 | 136 | |
AK157118.1 | - | 3 - 4 | 331 | 135 | |
CT010194.1 | - | 3 - 4 | 274 | 135 |