Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Wiz
Entrez Gene ID
22404
Gene Name
widely-interspaced zinc finger motifs
Gene Aliases
-
Chromosome Location
Chr.17: 32354047 - 32389439 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011717.4 NP_035847.2 6 - 7 2494 77
NM_212438.3 NP_997603.3 5 - 6 2150 77
NR_033525.1 8 - 9 2574 77
GenBank mRNA AB012265.1 - 10 - 11 4679 77
AB012266.1 - 6 - 7 2188 77
AB255388.1 - 5 - 6 2092 77
BC053035.1 - 8 - 9 2526 77

View Full Product Details