Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Grin2a
- Entrez Gene ID
-
14811
- Gene Name
- glutamate receptor, ionotropic, NMDA2A (epsilon 1)
- Gene Aliases
-
GluN2A, NMDAR2A, NR2A
- Chromosome Location
-
Chr.16: 9573737 - 9996500 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK086290.1 |
- |
1 - 2 |
214 |
55 |
View Full Product Details