The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181596.4 | NP_853627.2 | 2 - 3 | 248 | 132 |
GenBank mRNA | AB086819.1 | - | 1 - 2 | 169 | 132 |
AJ514933.1 | - | 1 - 2 | 172 | 132 | |
AJ536019.1 | - | 2 - 3 | 211 | 132 | |
AK158401.1 | - | 2 - 3 | 212 | 132 | |
BC117106.1 | - | 1 - 2 | 161 | 132 | |
DY246276.1 | - | 2 - 3 | 280 | 132 |