Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Abhd5
- Entrez Gene ID
-
67469
- Gene Name
- abhydrolase domain containing 5
- Gene Aliases
-
1300003D03Rik, 2010002J10Rik, CDS, CGI-58, IECN5, NCIE2
- Chromosome Location
-
Chr.9: 122351539 - 122381524 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK019488.2 |
- |
1 - 2 |
105 |
101 |
View Full Product Details