The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007570.2 | NP_001007571.1 | 7 - 8 | 734 | 94 |
GenBank mRNA | AK143301.1 | - | 6 - 7 | 1196 | 94 |
BC022213.1 | - | 7 - 8 | 724 | 94 | |
BC025937.1 | - | 7 - 8 | 709 | 94 | |
BC036140.1 | - | 7 - 8 | 713 | 94 |