Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Haus8
Entrez Gene ID
76478
Gene Name
4HAUS augmin-like complex, subunit 8
Gene Aliases
2410004L22Rik, Hice1
Chromosome Location
Chr.8: 71251124 - 71272590 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163042.1 NP_001156514.1 4 - 5 491 72
GenBank mRNA AK161850.1 - - 363 72
BC003935.1 - 4 - 5 259 72
DV651534.1 - 4 - 5 398 72

View Full Product Details