The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016785.2 | NP_058065.2 | 7 - 8 | 735 | 227 |
GenBank mRNA | AF037043.1 | - | 7 - 8 | 644 | 227 |
AF037044.1 | - | 6 - 7 | 595 | 227 | |
AF037045.1 | - | 6 - 7 | 595 | 227 | |
AF046887.1 | - | 7 - 8 | 721 | 227 | |
AK002335.1 | - | 7 - 8 | 642 | 122 | |
BC021598.1 | - | 6 - 7 | 622 | 225 |