The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166463.1 | NP_001159935.1 | 5 - 6 | 616 | 60 |
NM_001166464.1 | NP_001159936.1 | 6 - 7 | 625 | 60 | |
NM_001166465.1 | NP_001159937.1 | 5 - 6 | 616 | 60 | |
NM_001166466.1 | NP_001159938.1 | 6 - 7 | 625 | 60 | |
NM_009262.3 | NP_033288.2 | 6 - 7 | 625 | 60 | |
GenBank mRNA | AK036189.1 | - | 6 - 7 | 626 | 60 |
AK039058.1 | - | 5 - 6 | 598 | 60 | |
AK049019.1 | - | 5 - 6 | 584 | 60 | |
AK051191.1 | - | 5 - 6 | 576 | 60 | |
BC075625.1 | - | 6 - 7 | 568 | 60 | |
BC158020.1 | - | 6 - 7 | 532 | 60 | |
X92864.1 | - | 6 - 7 | 617 | 60 |