Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dnajc25
Entrez Gene ID
72429
Gene Name
DnaJ (Hsp40) homolog, subfamily C, member 25
Gene Aliases
2010109C08Rik, 2010203O07Rik
Chromosome Location
Chr.4: 58994810 - 59023398 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033165.3 NP_001028337.2 1 - 2 367 69
GenBank mRNA AK136359.1 - 1 - 2 61 69

View Full Product Details